VWR®, Shoe Covers, CPE
Supplier: VWR Collection
Über diesen Artikel
Basic shoe covers manufactured from a low-density chlorinated polyethylene fabric, which is designed to protect against dirt, grime, and liquid splash.
- CPE is water resistant and provides good resistance against chemicals
- Economical and lint-free
Ideal for laboratory use.
* One size fits all
2 mögliche Produktvariationen vorhanden
Product Details & Documents
Basic shoe covers manufactured from a low-density chlorinated polyethylene fabric, which is designed to protect against dirt, grime, and liquid splash.
- CPE is water resistant and provides good resistance against chemicals
- Economical and lint-free
Ideal for laboratory use.
* One size fits all
Spezifikationen für diese Produktfamilie
Spezifikationen:
- Catalog No:
- 113-1491
- 113-1492
- Garment application:
- Laboratory
- Laboratory
Spezifikationen:
Product Family Options
Produktinformationen
- ColourShoe sizeAvailabilityPrice
- Spezifikationen:
More
Specifications
Cat. No.113-1491Garment applicationLaboratory - Spezifikationen:
More
Specifications
Cat. No.113-1492Garment applicationLaboratory