Gel DNA kit, SpinPrep™
Catalog # 70852-3
Supplier: Merck Millipore (Novagen)
CAS Number:
Specifications
- Pk:100 Tests
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:DNA gel bands
- Sample type:Gel matrix
- Target molecule:DNA
- Description:SpinPrep™ Gel DNA Kit
- Description:SpinPrep™ Gel DNA Kit
Specifications
About this item
The SpinPrep™ Gel DNA Kit enables efficient extraction of DNA fragments of 150 bp to >12000 bp in size from agarose gels. The procedure uses GelMelt™ solution to dissolve the gel slice, followed by adsorption of the DNA to a silica membrane in a spin column format. After a wash step, the purified DNA is eluted in low salt buffer. Each spin column can bind up to 20 µg DNA. Recoveries are consistently in the range of 50 to 90% under standard conditions.
- No organic extraction or alcohol precipitation
- Total preparation time less than 30 minutes
- No low melting point agarose is required
- Receiver tubes (1,5 ml) for the final DNA elution step are included