Order Entry
Germany
ContactUsLinkComponent
 
Water detection paste N, Supelco®
Catalog # 1.08641.0001
Supplier:  Merck
CAS Number:  
undefined
Water detection paste N, Supelco®
Catalog # 1.08641.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    150 g
  • Description:
    Water detection paste N 1
  • M-Clarity quality level:
    MQ100

Specifications

About this item

Typically used to determine whether containers filled with Liquids (not miscible with water) such as engine fuels and oils that may contain water below the oil Liquid Layer. The paste is applied to any suitable dipstick, which is then inserted into the container. The paste changes colour on exposure to water.

  • Spec. density at 20 °C approx. 1,6 g/cm3
  • pH (HO) approx. 11
  • Suitability for the detection of water