Order Entry
Germany
ContactUsLinkComponent
 
Corning® Human fibronectin (from plasma)
 :  Corning
    
 
undefined
Corning® Human fibronectin (from plasma)
 :  Corning
    
 

 

  • Source:
    Human plasma
  • Species:
    Human
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Storage conditions:
    Stable for three months at 2 to 8 °C (lyophilised) or two weeks at −20 °C (solubilised). Do not store in frost-free freezer. Avoid multiple freeze-thaws.
  • Environmental certification:
    ISO 14001 - Bedford
  • Protein/peptide name:
    Fibronectin
  • Formulation:
    Lyophilised from CAPS buffer containing NaCl and CaCl₂, pH 11,0; reconstitute in distilled water (do not agitate or swirl)