Order Entry
Germany
ContactUsLinkComponent
 
Corning® MITO+ Serum Extender, Corning
  734-1317
 :  Corning
   Low Manufacturing Impact
 :  
Corning® MITO+ Serum Extender, Corning
  734-1317
 :  Corning
   Low Manufacturing Impact
 :  

 

  • Pk:
    5 mL
  • Description:
    Corning® MITO+ Serum Extender
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 - Bedford
  • Formulation:
    Lyophilised from a solution of Dulbecco's Phosphate Buffered Saline (DPBS) containing ECGS, EGF, insulin, human transferrin, triiodotryronine, progesterone, oestradiol-17ß, testosterone, hydrocortisone, selenous acid, and o-phosphorylethanolamine; reconstitute in 5 ml distilled H₂O (stock solution).
  • Quality control:
    Tested for ability to promote proliferation of BALB/c-3T3 cells in serum-reduced medium
    Filtered (0,2 µm membrane) and tested and found negative for bacteria, fungi and mycoplasma
  • Recommended concentration:
    1:1000 (dilute stock solution in serum-reduced medium)
  • Storage and stability:
    Stable for at least three months at 2 to 8 °C (lyophilised) or 3 months at −20 °C (solubilised).  Avoid multiple freeze-thaw cycles.

 

 

MITO+ Serum Extender is a concentrated, fully defined formulation of hormones, growth factors (EGF and FGF), and other metabolites (insulin and steriod hormones).

It can be used to culture a variety of cells under serum-free or serum-reduced conditions.

Tested for ability to promote proliferation of BALB/c-3T3 cells in serum-reduced medium. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi and mycoplasma.

(5ml; 5L equivalent).