TLC/HPTLC sprayers
TLC/HPTLC sprayer
Catalog # CAMA022.6530
Supplier: CAMAG
Jump to:
Product Details & Documents
Reagents are atomised into a fine aerosol spray with particles in the range of 0,3 to 10 µm.
- Function is electro-pneumatic
- Homogeneous distribution over the layer at a low reagent consumption
: Complete sprayer consists of charger and a pump unit with two kinds of spray heads, type A for spray solutions of normal viscosity (organic solvents), and type B for liquids of higher viscosity.
Specifications
- Description:TLC/HPTLC sprayer
- Seq:01
Accessories
Product Information
- Catalog #AvailabilityPrice
- Spray heads, type A and type B
Product Information
- Catalog #AvailabilityPrice
- Reagent bottle with cap, 100 ml
Product Information
- Catalog #AvailabilityPrice
- Spray heads, type B
Product Information
- Catalog #AvailabilityPrice
- Reagent bottle with cap, 50 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



