Orders LinkContactUsLinkComponent
Cassettes for multi-channel pump heads
Cassettes for multi-channel pump heads
Certificates
Cassettes for multi-channel pump heads
Cassettes for multi-channel pump heads
4 Options Available Below
Product Details & Documents
The cassettes can be replaced while the device is pumping. The contact pressure of the rollers is adjusted by means of an adjusting screw. Tubes of various materials and sizes can be inserted in any cassette.
Cassette small
Rated flow 0,005 to 40,8 ml/min
For tubing with 0,9 mm wall thickness
For tubing Ø 0,2/0,5/0,9/1,4 and 2,8 mm
For three stopper tubing
Cassette medium
Rated flow 0,29 to 25,2 ml/min
For tubing with 1,6 mm wall thickness
For tubing Ø 0,8 and 1,7 mm
For standard tubing
Cassette large
Rated flow 1,0 to 336 ml/min
For tubing with 1,6 mm wall thickness
For tubing Ø 1,7/3,1/4,8 and 6,4 mm
For standard tubing
Multi-channel adapter
For fitting and converting tube pump drives PD 5001, PD 5101, and PD 5201 for use as multi-channel pumps.
Product Family Options
Product Information
  • Availability
    Price
  • 224-1453
    Cassette large
    Cassette large
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 305,00 €
    305,00 €
    1 * 1 ST
       305,00 €
  • 224-1452
    Cassette medium
    Cassette medium
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 271,00 €
    271,00 €
    1 * 1 ST
       271,00 €
  • 224-1451
    Cassette small
    Cassette small
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 269,00 €
    269,00 €
    1 * 1 ST
       269,00 €
  • 224-1349
    Multi-channel adaptor
    Multi-channel adaptor
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 140,00 €
    140,00 €
    1 * 1 ST
       140,00 €