Orders LinkContactUsLinkComponent
Accurate probes
 :   Testo
Accurate probes
 :  Testo
Accurate probes
Accurate probes
Product Family Options
  • Description
    Range
    Accuracy
    Application
    Ø×L
  • NTC stainless steel probe with PTFE cable, cable length 1,5 m, probe shaft tip Ø×L: 3×15 mm
    −50...+150 °C
    ±0,5 % of mv (+100...+150 °C)
    ±0,2 °C (‒25...+74,9 °C)
    ±0,4 °C
    Food
    4×125 mm
    1 * 1 ST
    1 * 1 ST - 149,00 €
    149,00 €
    1 * 1 ST
       149,00 €
  • NTC waterproof flat surface probe, cable length 1,2 m, probe shaft tip Ø×L: 6×50 mm
    −50...+150 °C
    ±0,5 % of mv (100...+150 °C)
    ±0,2 °C (‒25...+74,9 °C)
    ±0,4 °C (Remaining range)
    Contact
    5×115 mm
    1 * 1 ST
    1 * 1 ST - 96,00 €
    96,00 €
    1 * 1 ST
       96,00 €
  • Robust NTC probe, cable length 1,1 m, probe shaft tip Ø×L: 4×50 mm
    −50...+125 °C
    ±0,2 °C (‒25...+80 °C)
    ±0,4 °C (Remaining range)
    Air
    5×115 mm
    1 * 1 ST
    1 * 1 ST - 87,00 €
    87,00 €
    1 * 1 ST
       87,00 €
  • Waterproof NTC probe, cable length 1,2 m, probe shaft tip Ø×L: 4×50 mm
    −50...+150 °C
    ±0,5 % of mv (100...+150 °C)
    ±0,2 °C (‒25...+74,9 °C)
    ±0,4 °C (remaining range)
    Penetration
    5×115 mm
    1 * 1 ST
    1 * 1 ST - 84,00 €
    84,00 €
    1 * 1 ST
       84,00 €