Headspace vials, WHEATON®
Supplier: DWK Life Sciences
Jump to:
About this item
These vials made of type I borosilicate glass feature rounded shoulders and bottoms, allowing for even heating and safer operation at higher pressures.
- Accepts 20 mm aluminium seal
- Top designed to provide a tight seal with septa
Ordering information: Aluminium seals must be ordered separately.
3 Options Available Below
Product Details & Documents
These vials made of type I borosilicate glass feature rounded shoulders and bottoms, allowing for even heating and safer operation at higher pressures.
- Accepts 20 mm aluminium seal
- Top designed to provide a tight seal with septa
Ordering information: Aluminium seals must be ordered separately.
Product Family Options
Flat bottom, beveled finish
Product Information
- CapacityØ×HMaterialColourCap includedAvailabilityPrice
Rounded bottom, beveled finish
Product Information
Aluminium seals
Product Information
- Catalog #SeptumØAvailabilityPrice