ContactUsLinkComponent
Microtome blades, Personna Plus®
Microtome blades, Personna Plus®
Catalog # PERS72-0114-0000
Supplier:  PERSONNA MEDICAL
Microtome blades, Personna Plus®
Catalog # PERS72-0114-0000
Supplier:  PERSONNA MEDICAL
Specifications
  • Description:
    Microtome blades
  • Material:
    Stainless steel
  • Seq:
    02
  • Pk:
    50
Specifications
About this item
Personna Plus® microtome blades with MicroCoat® coating.
  • MicroCoat® coating allows blades to glide with smooth and clean precision
  • Uniform ribbon lacking any grooves or split sections of paraffin
  • Transparent viewing area enabling the user to see the number of blades
  • Disposal reservoir prevents used blades from dropping out
Delivery: In a plastic safety dispenser.