Orders LinkContactUsLinkComponent
Staining dishes, WHEATON®
Staining dishes, WHEATON®
Supplier:   DWK Life Sciences
Staining dishes, WHEATON®
Supplier:  DWK Life Sciences
Certificates
Staining dishes, WHEATON®
Staining dishes, WHEATON®
6 Options Available Below
Product Details & Documents
Soda-lime glass and stainless steel.
  • Removable glass slide rack has an open bottom to facilitate rapid immersion and drainage, reducing carry-over
Standard for manual staining procedures. The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back to back of standard size 75×25 mm, 75×38 mm and 75×51 mm.
Documents
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • 631-2501
    Staining dish, only
    Staining dish, only
    95×76×64 mm
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 213,00 €
    213,00 €
    1 * 3 ST
       213,00 €
  • 631-2500
    Staining dish, complete, includes dish, cover and rack
    Staining dish, complete, includes dish, cover and rack
    95×76×64 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 1 070,00 €
    1 070,00 €
    1 * 6 ST
       1 070,00 €
  • 631-2502
    Cover for staining dish
    Cover for staining dish
    - mm
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 135,00 €
    135,00 €
    1 * 3 ST
       135,00 €
  • 631-2503
    Staining dish with cover
    Staining dish with cover
    95×76×64 mm
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 349,00 €
    349,00 €
    1 * 3 ST
       349,00 €
  • 631-2504
    Staining rack, only, for 10 to 20 slides
    Staining rack, only, for 10 to 20 slides
    - mm
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 204,00 €
    204,00 €
    1 * 3 ST
       204,00 €
  • 631-2505
    Handles for staining racks
    Handles for staining racks
    - mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 86,70 €
    86,70 €
    1 * 6 ST
       86,70 €