ContactUsLinkComponent
Complete sampler, Wheaton® E-Z
Complete sampler, Wheaton® E-Z
Catalog # WHEA885020
Supplier:  DWK Life Sciences
undefined
Complete sampler, Wheaton® E-Z
Catalog # WHEA885020
Supplier:  DWK Life Sciences
Specifications
  • Capacity:
    125 ml
  • Description:
    Complete sampler
  • Material:
    PP
  • Pk:
    1
Specifications
About this item
The filling ports are located above the bottle to ensure that it is completely filled to reduce the possibility of air entrapment.
  • Head/shaft assembly and borosilicate glass bottle with lined cap
  • Constructed of chemically resistant PP
  • Bottle with a GPI 33-430 thread
This sampler is an efficient and time saving method of sampling from drums and tanks.