Orders LinkContactUsLinkComponent
Drum thieves, Wheaton®
Drum thieves, Wheaton®
Supplier:   DWK Life Sciences
Drum thieves, Wheaton®
Supplier:  DWK Life Sciences
Certificates
Drum thieves, Wheaton®
Drum thieves, Wheaton®
3 Options Available Below
Product Details & Documents
Drum thieves made of soda lime glass for quick disposal.
  • Conventional open end drum thieves
  • Just insert empty tube in drum
  • Hold thumb over top and withdraw
Product Family Options
Product Information
  • Material
    Capacity
    Availability
    Price
  • WHEA885300
    Drum thieves
    Drum thieves
    Glass
    25 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 484,00 €
    484,00 €
    1 * 100 ST
       484,00 €
  • WHEA885302
    Drum thieves
    Drum thieves
    Glass
    75 ml
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 200,00 €
    200,00 €
    1 * 24 ST
       200,00 €
  • WHEA885303
    Drum thieves
    Drum thieves
    Glass
    150 ml
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 287,00 €
    287,00 €
    1 * 24 ST
       287,00 €