Order Entry
Germany
Orders LinkContactUsLinkComponent
Sieve rack

Sieve rack

Sieve rack, for max. 10 sieves to 203 mm Ø, 50 mm height
Catalog # 510-1991
Supplier:   Retsch GmbH
Sieve rack
Catalog # 510-1991
Supplier:  Retsch GmbH
Sieve rack
Sieve rack

Sieve rack, for max. 10 sieves to 203 mm Ø, 50 mm height

Price
322,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Holds a maximum of 10 sieves with diameters up to 203 mm.

Documents

Specifications
  • Description:
    Sieve rack, for max. 10 sieves to 203 mm Ø, 50 mm height
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.