VWR®, Crucibles, PTFE
Supplier: VWR Collection
Certificates
About this item
Isostatically moulded in pure PTFE, FDA approved.
- Chemically resistant and inert
- Crack-free
- Withstand maximum temperature of 280 °C
Isostatically moulded in pure PTFE, FDA approved.