Order Entry
Germany
ContactUsLinkComponent
Protein A Mag Sepharose Xtra
Protein A Mag Sepharose Xtra
Supplier:  Cytiva
Certificates
undefined
Protein A Mag Sepharose Xtra
Supplier:  Cytiva

Specifications

  • Catalog No:
  • 28-9670-56
  • 28-9670-62
  • Binding capacity:
  • >27 mg human IgG/ml medium
  • >27 mg human IgG/ml medium
  • Matrix:
  • Highly cross-linked spherical agarose including magnetite
  • Highly cross-linked spherical agarose including magnetite
  • Operating temperature:
  • Ambient
  • Ambient
  • Particle size:
  • 37 - 100 µm
  • 37 - 100 µm
  • Storage conditions:
  • Storage solution ethanol 20%, 10% medium slurry
  • Storage solution ethanol 20%, 10% medium slurry

Specifications

Magnetic separation racks

Product Information