Earmuffs, Peltor™ Optime™ I
Supplier: 3M
Certificates
About this item
The lightweight Peltor™ Optime™ I earmuffs provide a moderate level of attenuation that meets the needs of the majority of industrial applications.
- Liquid-sealing rings for improved comfort
- Large space inside cups helps reduce moisture and heat build-up
- Soft, wide cushions reduce pressure around the ears
- Easy to replace cushions and inserts help keep them hygienically clean
- Helmet-mounted version fits directly to many safety helmets without the need for an adapter
For industrial applications, such as workshops, sheet metal shops and printing works. Besides that, these low-profile earmuffs are also suitable for outdoor tasks such as lawn mowing or in connection with hobby and leisure activities.
SNR: 26 dB (neckband and helmet-mounted)
SNR: 27 dB (headband)
SNR: 28 dB (foldable)
Certifications: EN 352-1 (headband, neckband and foldable version); EN 352-3 (helmet-mounted version).