ContactUsLinkComponent
Dispenser, Measure-Matic™, SP Bel-Art
  BELAF116540000
undefined
Dispenser, Measure-Matic™, SP Bel-Art
  BELAF116540000
  • Volume:
    5 - 25 ml
  • Pk:
    1
Bottles, LDPE, translucent, for quick dispensing, with polypropylene measuring cup. Measuring cup slides onto a polypropylene delivery tube. The height of the tube determines the volume of liquid dispensed into the measuring cup when the bottle is squeezed.
  • Repetitive dispensing of the same volume of liquid
  • When the bottle pressure is released excess flluid returns into the bottle for spill free pouring
  • Good chemical resistance