VWR®, Magnetic Stirring Bar Retriever
Supplier: VWR Collection
Certificates
About this item
PTFE covered, rare earth magnetic core. For the removal of stirring magnets from vessels of all kinds.
- Very good chemical resistance
- Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)