Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Stirrers
Stirring Rods
VWR®, Stirring Rods
VWR®, Stirring Rods
VWR®, Stirring Rods
Supplier:
VWR Collection
Certificates
Expand image
VWR®, Stirring Rods
Supplier:
VWR Collection
About this item
PTFE with steel core.
Inert and will not scratch, can be used up to 280 °C