Orders LinkContactUsLinkComponent
Insulated bags, silverBag
Supplier:   DELTA T
Insulated bags, silverBag
Supplier:  DELTA T
Certificates
Insulated bags, silverBag
Insulated bags, silverBag
6 Options Available Below
Product Details & Documents
SilverBags are the economical solution to the problem of short-term insulated transport of TempShells. The SilverBag range is made from tough nylon combined with an insulating material. The bags are designed so that the TempShells elements fit in the bag perfectly so that no space is wasted.
SilverBag 16 l: For holding up to five pairs of TempShells
Product Family Options
Product Information
  • Capacity
    Dimensions
    Weight
    Availability
    Price
  • 478-9919
    Bag with TempShells −30 °C and three FrameShells
    Bag with TempShells −30 °C and three FrameShells
    10 l
    310×180×260 mm
    500 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 547,00 €
    547,00 €
    1 * 1 ST
       547,00 €
  • 478-9920
    Bag with TempShells 4 °C and three FrameShells
    Bag with TempShells 4 °C and three FrameShells
    10 l
    310×180×260 mm
    500 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 493,00 €
    493,00 €
    1 * 1 ST
       493,00 €
  • 478-9921
    Bag with TempShells 22 °C and three FrameShells
    Bag with TempShells 22 °C and three FrameShells
    10 l
    310×180×260 mm
    500 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 478-9922
    Bag with TempShells −30 °C and four FrameShells
    Bag with TempShells −30 °C and four FrameShells
    16 l
    380×200×300 mm
    930 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 123,00 €
    123,00 €
    1 * 1 ST
       123,00 €
  • 478-9923
    Bag with TempShells 4 °C and four FrameShells
    Bag with TempShells 4 °C and four FrameShells
    16 l
    380×200×300 mm
    930 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 478-9924
    Bag with TempShells 22 °C and four FrameShells
    Bag with TempShells 22 °C and four FrameShells
    16 l
    380×200×300 mm
    930 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST