Absorbent powder, Chemizorb®
: Merck
For absorption of acids, alkalis or solvents. Thanks to its large surface area, the powder absorbs a very large amount of liquid. Low viscosity liquids are absorbed instantaneously, highly viscous media (glycerine, oils) within a few minutes.
- Grain size less than 0,1 mm (85%)
- Bulk density of 120 kg/m³
1 kilogram absorbs: 2,5 to 4,5 litres of aqueous solutions, acids, alkalis; 3 to 4 litres of organic solvent, benzene; 7 litres of viscous oils.