VWR® Traceable® Ultra, Digital Thermometers
These thermometers fit into flasks, cuvettes, test tubes and beakers for temperature readings in food, gases, samples, reagents, water baths, ovens, incubators and semi-solids. The long stem allows user to make measurements while stirring solutions in deep vessels. Stainless steel probe (Ø: 3,5 mm).
- Probe resistant to most chemicals
- Readings in °C updated every second
- Safe: Almost unbreakable plastic and stainless steel
Weight: < 28 g
: Comply with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
: Supplied with 1 year battery, Traceable® certificate of calibration and protective sleeve.
- Catalog No:
- 620-2080
- 620-2079
- 620-2081
- Model:
- 620-2080
- 620-2079
- 620-2081
- Probe:
- 289×3 mm
- 203×3 mm
- 289×3 mm
- Display:
- 6.35 mm digit height
- 6.35 mm digit height
- 6.35 mm digit height