Boron standard solution, concentrate, 1,000 mg B in water standard for AAS and ICP
Catalog # CPIIS4400-100074
Supplier: CPI
CAS Number:
Specifications
- Concentration:1000 mg
- Matrix:H₂O
- Pack type:Glass bottle
- Pk:100 mL
- Element:Boron
- Symbol:B