Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Quinazoline
Quinazoline
Quinazoline
Quinazoline
Supplier:
CHEMPUR
Certificates
Expand image
Quinazoline
Supplier:
CHEMPUR
About this item
CAS Number:
253-82-7
:
Formula:
C₈H₆N₂
MDL Number:
MFCD00006712
:
130.15 g/mol
EINECS:
205-965-3