Order Entry
Germany
ContactUsLinkComponent
Bismut(III) gallate basic hydrate ≥98% (by complexometric analysis) USP
Bismut(III) gallate basic hydrate ≥98% (by complexometric analysis) USP
Catalog # MOLE33641704-100G
Supplier:  Molekula
CAS Number:  
Bismut(III) gallate basic hydrate ≥98% (by complexometric analysis) USP
Catalog # MOLE33641704-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    98.0% Min (KT)
  • As:
    7.5ppm Max
  • Free Glallic Acid:
    0.5% Max
  • Alkalies & Alkaline Earth:
    0.5% Max
  • Loss on Drying:
    7.0% Max

Specification Test Results

About this item

 :
 :394.09 g/mol (anhy)
EINECS:202-742-2
CAS Number:99-26-3
Formula:C₇H₅BiO₆·xH₂O
MDL Number:MFCD01632692