5-(4-(Dimethylamino)benzylidene)rhodanine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:536-17-4
:MFCD04969049
Formula:C₁₂H₁₂N₂OS₂
MDL Number:MFCD00064857
:264.37 g/mol
:M-Clarity™ quality level = MQ100
EINECS:208-625-2