Order Entry
Germany
ContactUsLinkComponent
5-(4-(Dimethylamino)benzylidene)rhodanine
5-(4-(Dimethylamino)benzylidene)rhodanine
Catalog # MOLE24067530-50G
Supplier:  Molekula
CAS Number:  
5-(4-(Dimethylamino)benzylidene)rhodanine
Catalog # MOLE24067530-50G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :264.37 g/mol
EINECS:208-625-2
CAS Number:536-17-4
 :MFCD04969049
Formula:C₁₂H₁₂N₂OS₂
MDL Number:MFCD00064857