Order Entry
Germany
ContactUsLinkComponent
5-(4-(Dimethylamino)benzylidene)rhodanine 99%
5-(4-(Dimethylamino)benzylidene)rhodanine 99%
Certificates
undefined
5-(4-(Dimethylamino)benzylidene)rhodanine 99%
Certificates

Specification Test Results

  • Appearance:
    Red Powder
  • Infrared spectrum:
    Conforms
  • Solubility:
    (0.03 % in acetone) Clear
  • Titration with TBAH:
    98.5 to 101.5 %
  • Suitability:
    (for Ag determination) Passes test

Specification Test Results

Related Information