9-Fluorenylmethyl chloroformate (Fmoc chloride)
Catalog # MOLEM11086183
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:258.7 g/mol
EINECS:249-313-6
UN:2923
ADR:8,II
CAS Number:28920-43-6
Formula:C₁₅H₁₁ClO₂
MDL Number:MFCD00001138