(S)-(+)-Thiazolidine-4-carboxylic acid
Catalog # BACE4027488.0005
Supplier: BACHEM BIOCHEMICA
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:133.17 g/mol
EINECS:256-240-3
CAS Number:45521-09-3
Formula:C₄H₇NO₂S
MDL Number:MFCD00137425