L(+)-Ascorbic acid 50 g/l in aqueous solution for photometric analysis of silicate
Catalog # KRAF21501.2100
Supplier: Bernd Kraft
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:176.13 g/mol
Storage Temperature:Ambient
EINECS:200-066-2
CAS Number:50-81-7
Formula:C₆H₈O₆
MDL Number:MFCD00064328