L(+)-Ascorbic acid 100 g/l in aqueous solution for photometric analysis of phosphate
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:50-81-7
:
Formula:C₆H₈O₆
MDL Number:MFCD00064328
:176.13 g/mol
Storage Temperature:Ambient
EINECS:200-066-2