Thiazolidine-2-carboxylic acid, Sigma-Aldrich®
Catalog # SIAL467995-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:133.169 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:65126-70-7
:16310-13-7
Formula:C₄H₇NO₂S
MDL Number:MFCD00068005