Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Thiazolidine-2-carboxylic acid
Thiazolidine-2-carboxylic acid
Thiazolidine-2-carboxylic acid
Thiazolidine-2-carboxylic acid
MOLEM37939444
:
Molekula
:
Thiazolidine-2-carboxylic acid
MOLEM37939444
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
133.169 g/mol
:
65126-70-7
:
16310-13-7
:
C₄H₇NO₂S
:
MFCD00068005