Thiazolidine-2-carboxylic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:65126-70-7
:16310-13-7
Formula:C₄H₇NO₂S
MDL Number:MFCD00068005
:133.169 g/mol
Melting Pt:∼ 184 °C (dec.)
:GHS07
Storage Temperature:Ambient