Order Entry
Germany
ContactUsLinkComponent
Disodium fumarate for synthesis, Sigma-Aldrich®
Disodium fumarate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Disodium fumarate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    228 g/l (25 °C)
  • Molar Mass:
    160.04 g/mol
  • pH:
    7 - 8 (50 g/l, H2O)
  • Synonyms:
    Fumaric acid disodium salt
  • Hill Formula:
    C4H2Na2O4
  • Chemical Formula:
    NaOOCCH=CHCOONa

Specification Test Results

Related Information