Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Disodium fumarate
Disodium fumarate ≥98%
Disodium fumarate ≥98%
Disodium fumarate ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Disodium fumarate ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
17013-01-3
:
Formula:
C₄H₂Na₂O₄
MDL Number:
MFCD00064567
:
160.04 g/mol
EINECS:
241-087-7
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAA11276
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation