Catechol violet indicator for metal titration, Fluka™
:115-41-3
:
:C₁₉H₁₄O₇S
:MFCD00005868
:386.38 g/mol
:185 °C
:Ambient
:204-088-3
- Change range:complying
- Indicator Solution 0,1 % H₂O:complying
- Suitable for metal definition:complying
- Absorption maximum:436 - 446 nm
- Absorbtion (1%/1cm):400 - 500