Order Entry
Germany
Orders LinkContactUsLinkComponent
Catechol violet

Catechol violet

Glass bottle for solids, 5 g
Catalog # MOLE21118006-5G
Marketsource
Supplier:   Molekula
Catechol violet
Catalog # MOLE21118006-5G
Supplier:  Molekula
Marketsource
CAS Number:  115-41-3
Catechol violet

Glass bottle for solids, 5 g

CAS Number:  115-41-3
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Pyrocatechol Violet
  • CAS Number:115-41-3
  • Formula:C₁₉H₁₄O₇S
  • MDL Number:MFCD00005868
  • Molecular Weight:386.38 g/mol
  • EINECS:204-088-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications