Order Entry
Germany
ContactUsLinkComponent
Curcumin, Sigma-Aldrich®
Curcumin, Sigma-Aldrich®
Catalog # SIALC7727-500MG
Supplier:  Merck
CAS Number:  
Curcumin, Sigma-Aldrich®
Catalog # SIALC7727-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :368.39 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:207-280-5
CAS Number:458-37-7
Formula:C₂₁H₂₀O₆
MDL Number:MFCD00008365