Boron standard solution, 1000 mg/L B in water 0,5 M ammonium hydroxide (from B(OH₃)) standard for ICP
Catalog # KRAF03857.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Boron
- Symbol:B
Specifications
About this item
Certifications: Traceable to NIST