Boron standard solution, 10000 mg/L B in water 1 M ammonium hydroxide (from B(OH₃)) standard for ICP
Catalog # KRAF18169.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:10000 mg/L
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Boron
- Symbol:B