1,3,5-Trimethylhexahydro-1,3,5-triazine, Sigma-Aldrich®
Catalog # SIAL254673-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:129.21 g/mol
EINECS:203-612-8
CAS Number:108-74-7
Formula:C₆H₁₅N₃
MDL Number:MFCD00006057