Ammonium tetrachlorocuprate dihydrate, Sigma-Aldrich®
: Merck
:10060-13-6
:
:H₈Cl₄CuN₂·2H₂O
:MFCD00150384
:277.46 g/mol
:M-Clarity™ quality level = MQ100
:239-690-5
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
ε-Caprolactam, Sigma-Aldrich®
: Merck
Sodium acetate trihydrate ≥98%, GPR RECTAPUR®
: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection