Ammonium-Nitrogen standard solution, 1 mg/L NH₄ (as N) in water (from (NH₄)₂SO₄) for photometric analysis of ammonium-nitrogen
Catalog # KRAF12638.1000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1 ppm
- Pack type:Plastic bottle
- Pk:250 mL
- Description:Ammonium-Nitrogen standard solution
- Contents:(NH₄)₂SO₄ in H₂O