sec-Butyllithium 1.4 mol/l in cyclohexane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:598-30-1
:
Formula:C₄H₉Li
MDL Number:MFCD00009323
:64.06 g/mol
ADR:4.2,I
:M-Clarity™ quality level = MQ200
EINECS:209-927-7
UN:3394
Frequently Bought Together