Bismuth standard solution, 1000 mg/L Bi in 1 M nitric acid (from Bi) standard for ICP
Catalog # KRAF03846.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:1 M HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Bismuth
- Symbol:Bi
Specifications
About this item
Certifications: Traceable to NIST.