Bismuth standard solution, 1000 mg/L Bi in 10% nitric acid (from Bi) standard for AAS and ICP
Catalog # JOBIPLBI4-2X
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:10% HNO₃
- Pack type:Glass bottle
- Pk:500 mL
- Element:Bismuth
- Symbol:Bi
Specifications
About this item
Certifications: Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.