Order Entry
Germany
ContactUsLinkComponent
Bismuth standard solution, 1,000 mg/L Bi in 10% nitric acid standard for ICP
Bismuth standard solution, 1,000 mg/L Bi in 10% nitric acid standard for ICP
Catalog # JOBIJYICP-BI
CAS Number:  
Bismuth standard solution, 1,000 mg/L Bi in 10% nitric acid standard for ICP
Catalog # JOBIJYICP-BI
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    10% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Bismuth
  • Symbol:
    Bi

Specifications

About this item